Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Fluoro-1-iodo-2-nitrobenzene, 97%

Click to view available options
Quantity:
1g
5g
364-77-2
C6H3FINO2
266.998
MFCD00153169
ANPFMDLUEWNKIM-UHFFFAOYSA-N
2-iodo-5-fluoronitrobenzene,5-fluoro-2-iodonitrobenzene,4-fluoro-2-nitroiodobenzene,1-fluoro-4-iodo-3-nitrobenzene,benzene, 4-fluoro-1-iodo-2-nitro,pubchem1058,acmc-209y4c,ksc495i0t,1-iodo-2-nitro-4-fluorobenzene,buttpark 29\07-56
2737347
4-fluoro-1-iodo-2-nitrobenzene
C1=CC(=C(C=C1F)[N+](=O)[O-])I
364-77-2
C6H3FINO2
266.998
MFCD00153169
ANPFMDLUEWNKIM-UHFFFAOYSA-N
2-iodo-5-fluoronitrobenzene,5-fluoro-2-iodonitrobenzene,4-fluoro-2-nitroiodobenzene,1-fluoro-4-iodo-3-nitrobenzene,benzene, 4-fluoro-1-iodo-2-nitro,pubchem1058,acmc-209y4c,ksc495i0t,1-iodo-2-nitro-4-fluorobenzene,buttpark 29\07-56
2737347
4-fluoro-1-iodo-2-nitrobenzene
C1=CC(=C(C=C1F)[N+](=O)[O-])I
Sensitivity Light sensitive
Refractive Index 1.615
Boiling Point 110°C (4mmHg)
Quantity 1g
Beilstein 3253380
Formula Weight 267
Percent Purity 97%
Chemical Name or Material 4-Fluoro-1-iodo-2-nitrobenzene

GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures