Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Fluoro-2-iodoaniline, 98%

Click to view available options
Quantity:
5g
25g
61272-76-2
C6H5FIN
237.016
MFCD00042584
SETOTRGVPANENO-UHFFFAOYSA-N
2-iodo-4-fluoroaniline,4-fluoro-2-indoaniline,4-fluoro-2-iodoanline,4-fluoro-2-iodo-aniline,4-fluoro-2-iodophenylamine,4-fluoro-2-iodo-phenylamine,benzenamine, 4-fluoro-2-iodo,zlchem 368,pubchem3413,2-iodo-4-fluoro-aniline
2778283
4-fluoro-2-iodoaniline
C1=CC(=C(C=C1F)I)N
61272-76-2
C6H5FIN
237.016
MFCD00042584
SETOTRGVPANENO-UHFFFAOYSA-N
2-iodo-4-fluoroaniline,4-fluoro-2-indoaniline,4-fluoro-2-iodoanline,4-fluoro-2-iodo-aniline,4-fluoro-2-iodophenylamine,4-fluoro-2-iodo-phenylamine,benzenamine, 4-fluoro-2-iodo,zlchem 368,pubchem3413,2-iodo-4-fluoro-aniline
2778283
4-fluoro-2-iodoaniline
C1=CC(=C(C=C1F)I)N
Applications
4-Fluoro-2-iodoaniline is used as an organic chemical synthesis intermediate.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Quantity 5g
UN Number UN2810
Formula Weight 237.02
Percent Purity 98%
Chemical Name or Material 4-Fluoro-2-iodoaniline

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.