Alfa Aesar™ 4-Fluoro-2-isopropoxybenzeneboronic acid, 97%

Manufacturer: Alfa Aesar™ H5533803

Quantity 1g

 View more versions of this product

Catalog No. AAH5533803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 480438-59-3
    Formula Weight 198
    MDL Number MFCD06411287
    Chemical Name or Material 4-Fluoro-2-isopropoxybenzeneboronic acid

    Specifications

    Formula Weight 198
    MDL Number MFCD06411287
    Physical Form Solid
    CAS 480438-59-3
    Quantity 1g
    Chemical Name or Material 4-Fluoro-2-isopropoxybenzeneboronic acid
    Assay 97%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit