Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Fluoro-2-methoxyphenol, 97%

Click to view available options
Quantity:
1g
5g
25g
450-93-1
C7H7FO2
142.129
MFCD00070797
OULGLTLTWBZBLO-UHFFFAOYSA-N
4-fluoroguaiacol,5-fluoro-2-hydroxyanisole,4-fluoro-2-methoxy-phenol,phenol, 4-fluoro-2-methoxy,4-fluoro-2-methyoxyphenol,pubchem3880,acmc-1agod,2-methoxy-4-fluorophenol,4-fluoro-2-methoxyphenol
2737368
4-fluoro-2-methoxyphenol
COC1=C(C=CC(=C1)F)O
450-93-1
C7H7FO2
142.129
MFCD00070797
OULGLTLTWBZBLO-UHFFFAOYSA-N
4-fluoroguaiacol,5-fluoro-2-hydroxyanisole,4-fluoro-2-methoxy-phenol,phenol, 4-fluoro-2-methoxy,4-fluoro-2-methyoxyphenol,pubchem3880,acmc-1agod,2-methoxy-4-fluorophenol,4-fluoro-2-methoxyphenol
2737368
4-fluoro-2-methoxyphenol
COC1=C(C=CC(=C1)F)O
Flash Point 102°C (216°F)
Odor Phenol-like
Boiling Point 195°C
Quantity 5g
Beilstein 5425200
Formula Weight 142.13
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2-methoxyphenol

GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P280-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.

Danger