Alfa Aesar™ 4-Fluoro-2-methoxyphenol, 97%

Manufacturer: Alfa Aesar™ B2221103

Quantity 1g

 View more versions of this product

Catalog No. AAB2221103

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 450-93-1
    Molecular Formula C7H7FO2
    Formula Weight 142.13
    MDL Number MFCD00070797
    Beilstein 5425200
    Synonym 4-Fluoroguaiacol
    Chemical Name or Material 4-Fluoro-2-methoxyphenol

    Specifications

    Formula Weight 142.13
    Flash Point 102°C (216°F)
    Odor Phenol-like
    Boiling Point 195°C
    Molecular Formula C7H7FO2
    MDL Number MFCD00070797
    CAS 450-93-1
    Beilstein 5425200
    Synonym 4-Fluoroguaiacol
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H318-H302-H335-H315
    Causes serious eye damage.
    Harmful if swallowed.
    May cause respiratory irritation.
    Causes skin irritation.
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF eye irritation persists: Get medical advice/attention.
    Quantity 1g
    Chemical Name or Material 4-Fluoro-2-methoxyphenol
    Assay 97%

    GHS H Statement
    H318-H302-H335-H315
    Causes serious eye damage.
    Harmful if swallowed.
    May cause respiratory irritation.
    Causes skin irritation.

    GHS P Statement
    P280-P305+P351+P338-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF eye irritation persists: Get medical advice/attention.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit