Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-2-methoxyphenylboronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
179899-07-1
C7H8BFO3
169.95
MFCD03788423
ADJBXDCXYMCCAD-UHFFFAOYSA-N
4-fluoro-2-methoxyphenyl boronic acid,2-methoxy-4-fluorophenylboronic acid,4-fluoro-2-methoxybenzeneboronic acid,4-fluoro-2-methoxy-phenyl boronic acid,4-fluoro-2-methoxyphenyl-boronic acid,5-fluoroanisole-2-boronic acid,4-fluoro-2-methoxy phenyl boronic acid,boronic acid, 4-fluoro-2-methoxyphenyl,pubchem19716,acmc-1c2tm
2782672
(4-fluoro-2-methoxyphenyl)boronic acid
B(C1=C(C=C(C=C1)F)OC)(O)O
179899-07-1
C7H8BFO3
169.95
MFCD03788423
ADJBXDCXYMCCAD-UHFFFAOYSA-N
4-fluoro-2-methoxyphenyl boronic acid,2-methoxy-4-fluorophenylboronic acid,4-fluoro-2-methoxybenzeneboronic acid,4-fluoro-2-methoxy-phenyl boronic acid,4-fluoro-2-methoxyphenyl-boronic acid,5-fluoroanisole-2-boronic acid,4-fluoro-2-methoxy phenyl boronic acid,boronic acid, 4-fluoro-2-methoxyphenyl,pubchem19716,acmc-1c2tm
2782672
(4-fluoro-2-methoxyphenyl)boronic acid
B(C1=C(C=C(C=C1)F)OC)(O)O
Quantity 5g
Melting Point 133.9°C to 137.9°C
Infrared Spectrum Authentic
Color White
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 169.95
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2-methoxyphenylboronic acid
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

GHS Signal Word: Warning