Alfa Aesar™ 4-Fluoro-2-methylaniline, 97%

Manufacturer: Alfa Aesar™ A1160906

Quantity 5g

 View more versions of this product

Catalog No. AAA1160906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 452-71-1
    Molecular Formula C7H8FN
    Formula Weight 125.15
    MDL Number MFCD00007832
    UN Number UN2810
    Beilstein 1931798
    Synonym 2-Amino-5-fluorotoluene; 4-Fluoro-o-toluidine
    Chemical Name or Material 4-Fluoro-2-methylaniline

    Specifications

    Formula Weight 125.15
    Health Hazard 2 GHS H Statement
    H311-H373-H227-H302-H332-H315-H319
    Toxic in contact with skin.
    May cause damage to organs through prolonged or repeated exposure.
    Combustible liquid.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P210-P260-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    MDL Number MFCD00007832
    UN Number UN2810
    Synonym 2-Amino-5-fluorotoluene; 4-Fluoro-o-toluidine
    Flash Point 87°C (188°F)
    Boiling Point 90° to 92°C (16mmHg)
    CAS 452-71-1
    Beilstein 1931798
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
    Molecular Formula C7H8FN
    RTECS Number CY0500000
    Odor Characteristic
    Quantity 5g
    Chemical Name or Material 4-Fluoro-2-methylaniline
    Assay 97%

    GHS H Statement
    H311-H373-H227-H302-H332-H315-H319
    Toxic in contact with skin.
    May cause damage to organs through prolonged or repeated exposure.
    Combustible liquid.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P210-P260-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit