Alfa Aesar™ 4-Fluoro-2-methylbenzaldehyde, 97%

Manufacturer: Alfa Aesar™ H2626609

Quantity 10g

 View more versions of this product

Catalog No. AAH2626609

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 63082-45-1
    Molecular Formula C8H7FO
    Formula Weight 138.14
    MDL Number MFCD02261764
    Synonym 4-Fluoro-o-tolualdehyde
    Chemical Name or Material 4-Fluoro-2-methylbenzaldehyde

    Specifications

    Formula Weight 138.14
    Flash Point 82°C (180°F)
    Molecular Formula C8H7FO
    MDL Number MFCD02261764
    CAS 63082-45-1
    Synonym 4-Fluoro-o-tolualdehyde
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 10g
    Chemical Name or Material 4-Fluoro-2-methylbenzaldehyde
    Assay 97%

    GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit