Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Fluoro-2-methylbenzeneboronic acid, 98%

Click to view available options
Quantity:
1g
5g
25g
139911-29-8
C7H8BFO2
153.947
MFCD02093072
IQMLIVUHMSIOQP-UHFFFAOYSA-N
2-methyl-4-fluorophenylboronic acid,4-fluoro-2-methylbenzeneboronic acid,4-fluoro-2-methylphenyl boronic acid,4-fluoro-2-methylphenyl boranediol,4-fluoro-2-methyl-phenyl boronic acid,boronic acid, 4-fluoro-2-methylphenyl,4-fluoro-2-methylphenylboronicacid,pubchem1852,acmc-209cjp
2734665
(4-fluoro-2-methylphenyl)boronic acid
B(C1=C(C=C(C=C1)F)C)(O)O
139911-29-8
C7H8BFO2
153.947
MFCD02093072
IQMLIVUHMSIOQP-UHFFFAOYSA-N
2-methyl-4-fluorophenylboronic acid,4-fluoro-2-methylbenzeneboronic acid,4-fluoro-2-methylphenyl boronic acid,4-fluoro-2-methylphenyl boranediol,4-fluoro-2-methyl-phenyl boronic acid,boronic acid, 4-fluoro-2-methylphenyl,4-fluoro-2-methylphenylboronicacid,pubchem1852,acmc-209cjp
2734665
(4-fluoro-2-methylphenyl)boronic acid
B(C1=C(C=C(C=C1)F)C)(O)O
Quantity 25g
Melting Point 199°C to 201°C
Formula Weight 153.95
Percent Purity 98%
Chemical Name or Material 4-Fluoro-2-methylbenzeneboronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning