Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-2-methylbenzenesulfonyl chloride, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
7079-48-3
C7H6ClFO2S
208.64
XLPGWKNCWMFHOD-UHFFFAOYSA-N
4-fluoro-2-methylbenzene-1-sulfonyl chloride,4-fluoro-2-methylbenzenesulfonylchloride,4-fluoro-2-methylbenzenesulphonyl chloride,4-fluoro-2-methyl-benzenesulfonyl chloride,chloro 4-fluoro-2-methylphenyl sulfone,pubchem11719,ablock ab-12-8862,4-fluoro-2-methylphenylsulfonyl chloride,4-fluoro-2-methylbenzene-1-sulfonylchloride,benzenesulfonylchloride, 4-fluoro-2-methyl
2778602
4-fluoro-2-methylbenzenesulfonyl chloride
CC1=C(C=CC(=C1)F)S(=O)(=O)Cl
7079-48-3
C7H6ClFO2S
208.64
XLPGWKNCWMFHOD-UHFFFAOYSA-N
4-fluoro-2-methylbenzene-1-sulfonyl chloride,4-fluoro-2-methylbenzenesulfonylchloride,4-fluoro-2-methylbenzenesulphonyl chloride,4-fluoro-2-methyl-benzenesulfonyl chloride,chloro 4-fluoro-2-methylphenyl sulfone,pubchem11719,ablock ab-12-8862,4-fluoro-2-methylphenylsulfonyl chloride,4-fluoro-2-methylbenzene-1-sulfonylchloride,benzenesulfonylchloride, 4-fluoro-2-methyl
2778602
4-fluoro-2-methylbenzenesulfonyl chloride
CC1=C(C=CC(=C1)F)S(=O)(=O)Cl
Packaging Glass Bottle
Boiling Point 150°C to 152°C (92.0 Torr)
Flash Point 110°C
Quantity 1g
Color Colorless to Yellow
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 208.64
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2-methylbenzenesulfonyl chloride
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger