Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-2-methylphenylboronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
139911-29-8
C7H8BFO2
153.95
MFCD02093072
IQMLIVUHMSIOQP-UHFFFAOYSA-N
2-methyl-4-fluorophenylboronic acid,4-fluoro-2-methylbenzeneboronic acid,4-fluoro-2-methylphenyl boronic acid,4-fluoro-2-methylphenyl boranediol,4-fluoro-2-methyl-phenyl boronic acid,boronic acid, 4-fluoro-2-methylphenyl,4-fluoro-2-methylphenylboronicacid,pubchem1852,acmc-209cjp
2734665
(4-fluoro-2-methylphenyl)boronic acid
B(C1=C(C=C(C=C1)F)C)(O)O
139911-29-8
C7H8BFO2
153.95
MFCD02093072
IQMLIVUHMSIOQP-UHFFFAOYSA-N
2-methyl-4-fluorophenylboronic acid,4-fluoro-2-methylbenzeneboronic acid,4-fluoro-2-methylphenyl boronic acid,4-fluoro-2-methylphenyl boranediol,4-fluoro-2-methyl-phenyl boronic acid,boronic acid, 4-fluoro-2-methylphenyl,4-fluoro-2-methylphenylboronicacid,pubchem1852,acmc-209cjp
2734665
(4-fluoro-2-methylphenyl)boronic acid
B(C1=C(C=C(C=C1)F)C)(O)O
Melting Point 194°C to 196°C
Color White
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 153.95
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2-methylphenylboronic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning