Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Fluoro-2-methylphenylmagnesium bromide, 0.50 M in 2-MeTHF, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
Description
It is an important raw material and intermediate used in organic synthesis, Pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis, Pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Soluble in 0.5 M in Tetrahydrofuran.
Notes
Store at 4°C. Air
It is an important raw material and intermediate used in organic synthesis, Pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Soluble in 0.5 M in Tetrahydrofuran.
Notes
Store at 4°C. Air
Specifications
Quantity | 100 mL |
UN Number | UN2924 |
Sensitivity | Air and moisture sensitive |
Solubility Information | Soluble in 0.5 M in Tetrahydrofuran. |
Formula Weight | 213.33 |
Concentration or Composition (by Analyte or Components) | 0.50M in 2-MeTHF |
Chemical Name or Material | 4-Fluoro-2-methylphenylmagnesium bromide |
Safety and Handling
missing translation for 'healthHazard1' | H225-H302-H314-H335 |
missing translation for 'healthHazard2' | GHS H Statement H225-H314 Highly flammable liquid and vapor. Causes severe skin burns and eye damage. |
missing translation for 'healthHazard3' | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S. |
TSCA | No |
Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'
Spot an opportunity for improvement?
missing translation for 'provideContentCorrection'