Alfa Aesar™ 4-Fluoro-2-n-propoxybenzeneboronic acid, 97%

Manufacturer: Alfa Aesar™ H55269MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH55269MD

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 480438-60-6
    Formula Weight 198
    MDL Number MFCD06411288
    Chemical Name or Material 4-Fluoro-2-n-propoxybenzeneboronic acid

    Specifications

    Formula Weight 198
    Physical Form Solid
    MDL Number MFCD06411288
    CAS 480438-60-6
    Quantity 250mg
    Chemical Name or Material 4-Fluoro-2-n-propoxybenzeneboronic acid
    Assay 97%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit