Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Fluoro-2-nitro-5-(trifluoromethyl)aniline, 98%

Click to view available options
Quantity:
250mg
1g
5g
179062-05-6
C7H4F4N2O2
224.115
MFCD03094267
WOSGMZUIHAFPSS-UHFFFAOYSA-N
5-amino-2-fluoro-4-nitrobenzotrifluoride,4-fluoro-2-nitro-5-trifluoromethyl aniline,benzenamine, 4-fluoro-2-nitro-5-trifluoromethyl,pubchem4933,ksc106o9t,2-fluoro-4-nitro-5-aminobenzotrifluoride,3-amino-6-fluoro-4-nitrobenzotrifluoride,4-fluoro-2-nitro-5-trifluoromethyl benzenamine,4-fluoro-2-nitro-5-trifluoromethyl phenylamine
2737686
4-fluoro-2-nitro-5-(trifluoromethyl)aniline
C1=C(C(=CC(=C1N)[N+](=O)[O-])F)C(F)(F)F
179062-05-6
C7H4F4N2O2
224.115
MFCD03094267
WOSGMZUIHAFPSS-UHFFFAOYSA-N
5-amino-2-fluoro-4-nitrobenzotrifluoride,4-fluoro-2-nitro-5-trifluoromethyl aniline,benzenamine, 4-fluoro-2-nitro-5-trifluoromethyl,pubchem4933,ksc106o9t,2-fluoro-4-nitro-5-aminobenzotrifluoride,3-amino-6-fluoro-4-nitrobenzotrifluoride,4-fluoro-2-nitro-5-trifluoromethyl benzenamine,4-fluoro-2-nitro-5-trifluoromethyl phenylamine
2737686
4-fluoro-2-nitro-5-(trifluoromethyl)aniline
C1=C(C(=CC(=C1N)[N+](=O)[O-])F)C(F)(F)F
Color White
Quantity 250mg
UN Number UN2811
Beilstein 14360199
Formula Weight 224.11
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 4-Fluoro-2-nitro-5-(trifluoromethyl)aniline

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.