Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Fluoro-2-nitrobenzoic acid, 98%

Click to view available options
Quantity:
1g
5g
25g
394-01-4
C7H4FNO4
185.11
MFCD00024300
YLUCXHMYRQUERW-UHFFFAOYSA-N
4-fluoro-2-nitrobenzoic acid,2-nitro-4-fluorobenzoic acid,4-fluoro-2-nitrobenzoicacid,benzoic acid, 4-fluoro-2-nitro,4-fluoro-2-nitro-benzoic acid,4-fluoro-2-nitrobenzoic acid 2-nitro-4-fluorobenzoic acid,pubchem1311,acmc-1aim5,ksc497m7t,2-carboxy-5-fluoronitrobenzene
2737416
C1=CC(=C(C=C1F)[N+](=O)[O-])C(=O)O
394-01-4
C7H4FNO4
185.11
MFCD00024300
YLUCXHMYRQUERW-UHFFFAOYSA-N
4-fluoro-2-nitrobenzoic acid,2-nitro-4-fluorobenzoic acid,4-fluoro-2-nitrobenzoicacid,benzoic acid, 4-fluoro-2-nitro,4-fluoro-2-nitro-benzoic acid,4-fluoro-2-nitrobenzoic acid 2-nitro-4-fluorobenzoic acid,pubchem1311,acmc-1aim5,ksc497m7t,2-carboxy-5-fluoronitrobenzene
2737416
C1=CC(=C(C=C1F)[N+](=O)[O-])C(=O)O
Melting Point 147°C to 150°C
Quantity 25g
Formula Weight 185.11
Percent Purity 98%
Chemical Name or Material 4-Fluoro-2-nitrobenzoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning