Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Fluoro-2-(trifluoromethyl)benzyl bromide, 97%

Click to view available options
Quantity:
1g
5g
206860-48-2
C8H5BrF4
257.026
MFCD00061176
JMNOONULDANZRZ-UHFFFAOYSA-N
4-fluoro-2-trifluoromethyl benzyl bromide,1-bromomethyl-4-fluoro-2-trifluoromethyl benzene,4-fluoro-2-trifluoromethyl benzylbromide,benzene, 1-bromomethyl-4-fluoro-2-trifluoromethyl,alpha-bromo-4-fluoro-2-trifluoromethyl toluene,1-bromomethyl-4-fluoro-2-trifluoromethyl-benzene,pubchem4642,acmc-20ai28,4-fluoro-2-trifluoromethylbenzyl bromide,4-fluoro-2-trifluoromethyl-benzyl bromide
2737576
1-(bromomethyl)-4-fluoro-2-(trifluoromethyl)benzene
C1=CC(=C(C=C1F)C(F)(F)F)CBr
206860-48-2
C8H5BrF4
257.026
MFCD00061176
JMNOONULDANZRZ-UHFFFAOYSA-N
4-fluoro-2-trifluoromethyl benzyl bromide,1-bromomethyl-4-fluoro-2-trifluoromethyl benzene,4-fluoro-2-trifluoromethyl benzylbromide,benzene, 1-bromomethyl-4-fluoro-2-trifluoromethyl,alpha-bromo-4-fluoro-2-trifluoromethyl toluene,1-bromomethyl-4-fluoro-2-trifluoromethyl-benzene,pubchem4642,acmc-20ai28,4-fluoro-2-trifluoromethylbenzyl bromide,4-fluoro-2-trifluoromethyl-benzyl bromide
2737576
1-(bromomethyl)-4-fluoro-2-(trifluoromethyl)benzene
C1=CC(=C(C=C1F)C(F)(F)F)CBr
Quantity 1g
Boiling Point 141°C
Flash Point 85°C (185°F)
UN Number UN1760
Formula Weight 257.03
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2-(trifluoromethyl)benzyl bromide

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.