Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Fluoro-2-(trifluoromethyl)cinnamic acid, 97+%

Click to view available options
Quantity:
1g
5g
243977-21-1
C10H6F4O2
234.15
MFCD00236297
OREHSCSLACWBTF-DUXPYHPUSA-N
4-fluoro-2-trifluoromethyl cinnamic acid,2e-3-4-fluoro-2-trifluoromethyl phenyl prop-2-enoic acid,3-4-fluoro-2-trifluoromethyl phenyl acrylic acid,pubchem8430,3-4-fluoro-2-trifluoromethyl-phenyl-acrylic acid,e-3-4-fluoro-2-trifluoromethyl phenyl acrylic acid,e-3-4-fluoro-2-trifluoromethyl phenyl-2-propenoic acid,e-3-4-fluoro-2-trifluoromethyl phenyl prop-2-enoic acid,e-3-4-fluoranyl-2-trifluoromethyl phenyl prop-2-enoic acid
5702896
(E)-3-[4-fluoro-2-(trifluoromethyl)phenyl]prop-2-enoic acid
C1=CC(=C(C=C1F)C(F)(F)F)C=CC(=O)O
243977-21-1
C10H6F4O2
234.15
MFCD00236297
OREHSCSLACWBTF-DUXPYHPUSA-N
4-fluoro-2-trifluoromethyl cinnamic acid,2e-3-4-fluoro-2-trifluoromethyl phenyl prop-2-enoic acid,3-4-fluoro-2-trifluoromethyl phenyl acrylic acid,pubchem8430,3-4-fluoro-2-trifluoromethyl-phenyl-acrylic acid,e-3-4-fluoro-2-trifluoromethyl phenyl acrylic acid,e-3-4-fluoro-2-trifluoromethyl phenyl-2-propenoic acid,e-3-4-fluoro-2-trifluoromethyl phenyl prop-2-enoic acid,e-3-4-fluoranyl-2-trifluoromethyl phenyl prop-2-enoic acid
5702896
(E)-3-[4-fluoro-2-(trifluoromethyl)phenyl]prop-2-enoic acid
C1=CC(=C(C=C1F)C(F)(F)F)C=CC(=O)O
Melting Point 181°C to 183°C
Quantity 5g
Formula Weight 234.15
Percent Purity ≥97%
Chemical Name or Material 4-Fluoro-2-(trifluoromethyl)cinnamic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning