Alfa Aesar™ 4-Fluoro-3,5-dimethylaniline, 97%

Manufacturer: Alfa Aesar™ H31663MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH31663MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1840-27-3
    Molecular Formula C8H10FN
    Formula Weight 139.17
    MDL Number MFCD09832247
    UN Number UN2811
    Synonym 4-Fluoro-3,5-xylidine
    Chemical Name or Material 4-Fluoro-3,5-dimethylaniline

    Specifications

    Formula Weight 139.17
    MDL Number MFCD09832247
    UN Number UN2811
    Melting Point 30° to 32°C
    Quantity 250mg
    Molecular Formula C8H10FN
    CAS 1840-27-3
    Synonym 4-Fluoro-3,5-xylidine
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H331-H373-H302-H312-H315-H319
    Toxic if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P260-P261-P280-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Chemical Name or Material 4-Fluoro-3,5-dimethylaniline
    Assay 97%

    Description

    Applications
    4-Fluoro-3,5-dimethylaniline is used in the novel synthesis of fluoroaromatics.

    Solubility
    Sparingly soluble in water 0.71 g/L @ 25°C.

    Notes
    Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

    GHS H Statement
    H331-H373-H302-H312-H315-H319
    Toxic if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P260-P261-P280-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit