Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Fluoro-3,5-dimethylaniline, 97%

Manufacturer:  Alfa Aesar™ H3166303

Quantity 1g

 View more versions of this product

Catalog No. AAH3166303


  Check Availability
Add to cart

Specifications

Description

Applications
4-Fluoro-3,5-dimethylaniline is used in the novel synthesis of fluoroaromatics.

Solubility
Sparingly soluble in water 0.71 g/L @ 25°C.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Description & Specifications

Chemical Identifiers

1840-27-3
139.17
UN2811
4-Fluoro-3,5-dimethylaniline
C8H10FN
MFCD09832247
4-Fluoro-3,5-xylidine

Specifications

30° to 32°C
1840-27-3
139.17
UN2811
4-Fluoro-3,5-dimethylaniline
1g
C8H10FN
MFCD09832247
4-Fluoro-3,5-xylidine
97%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H331-H373-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P260-P261-P280-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit