Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-3-hydroxybenzoic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
51446-31-2
C7H5FO3
156.11
QATKOZUHTGAWMG-UHFFFAOYSA-N
benzoic acid, 4-fluoro-3-hydroxy,4-fluoro-3-hydroxybenzoicacid,4-fluoro-3-hydroxy-benzoic acid,3-hydroxy-4-fluorobenzoic acid,pubchem3494,ksc497o4f,3-hydroxy-4-fluoro-benzoic acid,4-fluoro-3-hydroxy benzoic acid,timtec-bb sbb008505,buttpark 34\01-10
593635
4-fluoro-3-hydroxybenzoic acid
C1=CC(=C(C=C1C(=O)O)O)F
51446-31-2
C7H5FO3
156.11
QATKOZUHTGAWMG-UHFFFAOYSA-N
benzoic acid, 4-fluoro-3-hydroxy,4-fluoro-3-hydroxybenzoicacid,4-fluoro-3-hydroxy-benzoic acid,3-hydroxy-4-fluorobenzoic acid,pubchem3494,ksc497o4f,3-hydroxy-4-fluoro-benzoic acid,4-fluoro-3-hydroxy benzoic acid,timtec-bb sbb008505,buttpark 34\01-10
593635
4-fluoro-3-hydroxybenzoic acid
C1=CC(=C(C=C1C(=O)O)O)F
Packaging Glass Bottle
Melting Point 215.0°C to 217.0°C
Quantity 1g
Infrared Spectrum Authentic
Color Brown to White
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 156.11
Physical Form Powder or Crystalline Powder
Percent Purity 97%
Chemical Name or Material 4-Fluoro-3-hydroxybenzoic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning