Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-3-(hydroxymethyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Click to view available options
Quantity:
200mg
1g
481681-02-1
C7H8BFO3
169.946
MFCD08235075
PWMOQHMTXJYUGE-UHFFFAOYSA-N
4-fluoro-3-hydroxymethyl phenyl boronic acid,4-fluoro-3-hydroxymethyl phenylboronic acid,4-fluoro-3-hydroxymethyl benzeneboronic acid,4-fluoro-3-hydroxymethylphenylboronic acid,4-fluoro-3-hydroxymethyl phenyl boronic acid,boronic acid, 4-fluoro-3-hydroxymethyl phenyl,boronic acid,b-4-fluoro-3-hydroxymethyl phenyl,pubchem23767,acmc-209kcj,ksc496e1t
21865588
[4-fluoro-3-(hydroxymethyl)phenyl]boronic acid
B(C1=CC(=C(C=C1)F)CO)(O)O
481681-02-1
C7H8BFO3
169.946
MFCD08235075
PWMOQHMTXJYUGE-UHFFFAOYSA-N
4-fluoro-3-hydroxymethyl phenyl boronic acid,4-fluoro-3-hydroxymethyl phenylboronic acid,4-fluoro-3-hydroxymethyl benzeneboronic acid,4-fluoro-3-hydroxymethylphenylboronic acid,4-fluoro-3-hydroxymethyl phenyl boronic acid,boronic acid, 4-fluoro-3-hydroxymethyl phenyl,boronic acid,b-4-fluoro-3-hydroxymethyl phenyl,pubchem23767,acmc-209kcj,ksc496e1t
21865588
[4-fluoro-3-(hydroxymethyl)phenyl]boronic acid
B(C1=CC(=C(C=C1)F)CO)(O)O
Quantity 1g
Color White-Yellow
Melting Point 126°C
Formula Weight 169.95
Physical Form Crystal-Powder at 20°C
Chemical Name or Material 4-Fluoro-3-(hydroxymethyl)phenylboronic Acid (contains varying amounts of Anhydride)