Alfa Aesar™ 4-Fluoro-3-methyl-5-(trifluoromethyl)phenol, 97%

Manufacturer: Alfa Aesar™ H33276MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH33276MD

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C8H6F4O
    Formula Weight 194.13
    MDL Number MFCD22201069
    UN Number UN3261
    Chemical Name or Material 4-Fluoro-3-methyl-5-(trifluoromethyl)phenol

    Specifications

    Formula Weight 194.13
    MDL Number MFCD22201069
    UN Number UN3261
    Health Hazard 2 GHS H Statement
    H301-H314-H318
    Toxic if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Quantity 250mg
    Molecular Formula C8H6F4O
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Chemical Name or Material 4-Fluoro-3-methyl-5-(trifluoromethyl)phenol
    Assay 97%

    GHS H Statement
    H301-H314-H318
    Toxic if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit