Alfa Aesar™ 4'-Fluoro-3'-methylacetophenone, 97%

Manufacturer: Alfa Aesar™ H3249903

Quantity 1g

 View more versions of this product

Catalog No. AAH3249903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 369-32-4
    Molecular Formula C9H9FO
    Formula Weight 152.17
    MDL Number MFCD04972879
    Chemical Name or Material 4'-Fluoro-3'-methylacetophenone

    Specifications

    Formula Weight 152.17
    Odor Characteristic
    Quantity 1g
    Molecular Formula C9H9FO
    MDL Number MFCD04972879
    CAS 369-32-4
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Chemical Name or Material 4'-Fluoro-3'-methylacetophenone
    Assay 97%

    Description

    Applications
    4 ft.-Fluoro-3 ft.-methylacetophenone is used as pharmaceutical intermediate, and an intermediate in organic synthesis.

    Solubility
    Insoluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents, strong acids, bases.

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit