Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Fluoro-3-methylaniline, 97%

Click to view available options
Quantity:
5g
25g
452-69-7
C7H8FN
125.146
MFCD00025294
NYMDPDNETOLVBS-UHFFFAOYSA-N
2-fluoro-5-aminotoluene,4-fluoro-m-toluidine,5-amino-2-fluorotoluene,4-fluoro-3-methylbenzenamine,4-fluoro-3-methylphenylamine,benzenamine, 4-fluoro-3-methyl,4-fluoro-3-methyl aniline,4-fluoro-3-methyl-aniline,4-fluor-3-methylanilin,pubchem1513
67981
4-fluoro-3-methylaniline
CC1=C(C=CC(=C1)N)F
452-69-7
C7H8FN
125.146
MFCD00025294
NYMDPDNETOLVBS-UHFFFAOYSA-N
2-fluoro-5-aminotoluene,4-fluoro-m-toluidine,5-amino-2-fluorotoluene,4-fluoro-3-methylbenzenamine,4-fluoro-3-methylphenylamine,benzenamine, 4-fluoro-3-methyl,4-fluoro-3-methyl aniline,4-fluoro-3-methyl-aniline,4-fluor-3-methylanilin,pubchem1513
67981
4-fluoro-3-methylaniline
CC1=C(C=CC(=C1)N)F
Refractive Index 1.537
Density 1.12
Quantity 5g
Melting Point 32°C to 36°C
Flash Point 95°C (203°F)
Odor Characteristic
Boiling Point 85°C to 86°C (9mmHg)
UN Number UN2811
Beilstein 2935561
Formula Weight 125.15
Percent Purity 97%
Chemical Name or Material 4-Fluoro-3-methylaniline
Show More Show Less

GHS H Statement
H311-H373-H302-H332-H315-H319
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c

H301+H311+H331-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: II

EINECSNumber : 207-207-7

TSCA : No

Recommended Storage : Ambient temperatures