Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 4-Fluoro-3-methylbenzaldehyde, 97%
Click to view available options
Quantity:
1g
5g
25g
Description
Applications
4-Fluoro-3-methylbenzaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff field.
Solubility
Slightly soluble in water.
Notes
Store in refrigerator at 4°C. Incompatible material is strong oxidizing agents.
4-Fluoro-3-methylbenzaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff field.
Solubility
Slightly soluble in water.
Notes
Store in refrigerator at 4°C. Incompatible material is strong oxidizing agents.
Specifications
Density | 1.15 |
Refractive Index | 1.522 |
Sensitivity | Air Sensitive |
Quantity | 1g |
Boiling Point | 208°C |
Beilstein | 4797794 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 138.14 |
Percent Purity | 97% |
Chemical Name or Material | 4-Fluoro-3-methylbenzaldehyde |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures