Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Fluoro-3-methylbenzaldehyde, 97%

Click to view available options
Quantity:
1g
5g
25g
135427-08-6
C8H7FO
138.141
MFCD01631431
NRFKZFFVTGGEQF-UHFFFAOYSA-N
4-fluoro-3-methyl benzaldehyde,benzaldehyde, 4-fluoro-3-methyl,3-methyl-4-fluorobenzaldehyde,4-fluoro-3-methyl-benzaldehyde,4-fluoro-3-methylbenzaldehyde,pubchem1457,acmc-1chue,ksc494g7n,pharmabridge p-3415,3-methyl-4-fluoro benzaldehyde
2734874
4-fluoro-3-methylbenzaldehyde
CC1=C(C=CC(=C1)C=O)F
135427-08-6
C8H7FO
138.141
MFCD01631431
NRFKZFFVTGGEQF-UHFFFAOYSA-N
4-fluoro-3-methyl benzaldehyde,benzaldehyde, 4-fluoro-3-methyl,3-methyl-4-fluorobenzaldehyde,4-fluoro-3-methyl-benzaldehyde,4-fluoro-3-methylbenzaldehyde,pubchem1457,acmc-1chue,ksc494g7n,pharmabridge p-3415,3-methyl-4-fluoro benzaldehyde
2734874
4-fluoro-3-methylbenzaldehyde
CC1=C(C=CC(=C1)C=O)F
Applications
4-Fluoro-3-methylbenzaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff field.

Solubility
Slightly soluble in water.

Notes
Store in refrigerator at 4°C. Incompatible material is strong oxidizing agents.
Quantity 25g
Boiling Point 208°C
Beilstein 4797794
Formula Weight 138.14
Percent Purity 97%
Chemical Name or Material 4-Fluoro-3-methylbenzaldehyde

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning