Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Fluoro-3-methylbenzoic acid, 97%

Click to view available options
Quantity:
1g
5g
403-15-6
C8H7FO2
154.14
MFCD01631424
VOCCEVKUXUIHOI-UHFFFAOYSA-N
4-fluoro-m-toluic acid,4-fluoro-3-methylbenzoicacid,3-methyl-4-fluorobenzoic acid,4-fluoro-3-methyl-benzoic acid,3-methyl-4-fluoro benzoic acid,benzoic acid, 4-fluoro-3-methyl,pubchem1397,acmc-1aii0,ksc494s2d,rarechem al bo 2279
242828
4-fluoro-3-methylbenzoic acid
CC1=C(C=CC(=C1)C(=O)O)F
403-15-6
C8H7FO2
154.14
MFCD01631424
VOCCEVKUXUIHOI-UHFFFAOYSA-N
4-fluoro-m-toluic acid,4-fluoro-3-methylbenzoicacid,3-methyl-4-fluorobenzoic acid,4-fluoro-3-methyl-benzoic acid,3-methyl-4-fluoro benzoic acid,benzoic acid, 4-fluoro-3-methyl,pubchem1397,acmc-1aii0,ksc494s2d,rarechem al bo 2279
242828
4-fluoro-3-methylbenzoic acid
CC1=C(C=CC(=C1)C(=O)O)F
Quantity 1g
Melting Point 166°C to 168°C
Beilstein 1936531
Formula Weight 154.14
Percent Purity 97%
Chemical Name or Material 4-Fluoro-3-methylbenzoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning