Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-3-methylbenzoyl Chloride, TCI America™

Click to view available options
Quantity:
5g
25g
455-84-5
C8H6ClFO
172.583
MFCD01631426
FMFLKTDRPXYUGP-UHFFFAOYSA-N
4-fluoro-3-methylbenzoylchloride,4-fluoro-m-toluoyl chloride,4-fluoro-3-methyl-benzoyl chloride,benzoyl chloride, 4-fluoro-3-methyl,acmc-1arwr,5-chlorocarbonyl-2-fluorotoluene,4-fluoranyl-3-methyl-benzoyl chloride,4-fluoro-3-methyl benzoyl chloride,4-fluoro-3-methyl-benzoic acid chloride,4-fluoro-3-methylbenzoyl chloride
2774589
4-fluoro-3-methylbenzoyl chloride
CC1=C(C=CC(=C1)C(=O)Cl)F
455-84-5
C8H6ClFO
172.583
MFCD01631426
FMFLKTDRPXYUGP-UHFFFAOYSA-N
4-fluoro-3-methylbenzoylchloride,4-fluoro-m-toluoyl chloride,4-fluoro-3-methyl-benzoyl chloride,benzoyl chloride, 4-fluoro-3-methyl,acmc-1arwr,5-chlorocarbonyl-2-fluorotoluene,4-fluoranyl-3-methyl-benzoyl chloride,4-fluoro-3-methyl benzoyl chloride,4-fluoro-3-methyl-benzoic acid chloride,4-fluoro-3-methylbenzoyl chloride
2774589
4-fluoro-3-methylbenzoyl chloride
CC1=C(C=CC(=C1)C(=O)Cl)F
Quantity 5g
Boiling Point 115°C
Color Yellow
UN Number 3265
Formula Weight 172.58
Physical Form Clear Liquid at 20°C
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 4-Fluoro-3-methylbenzoyl Chloride

TSCA : No

Recommended Storage : Refrigerator