Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Fluoro-3-nitroaniline, 98%

Click to view available options
Quantity:
25g
100g
500g
364-76-1
C6H5FN2O2
156.116
MFCD00007833
LLIOADBCFIXIEU-UHFFFAOYSA-N
3-nitro-4-fluoroaniline,4-fluoro-3-nitrobenzenamine,benzenamine, 4-fluoro-3-nitro,aniline, 4-fluoro-3-nitro,4-fluoro-3-nitro-aniline,5-amino-2-fluoronitrobenzene,4-fluoro-3-nitro-phenylamine,3-nitro-4-fluoroanailine,3-nitro-4-fluoro aniline,4-fluoro-3-nitro aniline
67768
CHEBI:48642
4-fluoro-3-nitroaniline
C1=CC(=C(C=C1N)[N+](=O)[O-])F
364-76-1
C6H5FN2O2
156.116
MFCD00007833
LLIOADBCFIXIEU-UHFFFAOYSA-N
3-nitro-4-fluoroaniline,4-fluoro-3-nitrobenzenamine,benzenamine, 4-fluoro-3-nitro,aniline, 4-fluoro-3-nitro,4-fluoro-3-nitro-aniline,5-amino-2-fluoronitrobenzene,4-fluoro-3-nitro-phenylamine,3-nitro-4-fluoroanailine,3-nitro-4-fluoro aniline,4-fluoro-3-nitro aniline
67768
CHEBI:48642
4-fluoro-3-nitroaniline
C1=CC(=C(C=C1N)[N+](=O)[O-])F
Applications
4-Fluoro-3-nitroaniline is used in the preparation of commercial hair dyes. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

Solubility
Insoluble in water.

Notes
Incompatible with Oxidizing agents, Acids, Acid chlorides, Acid anhydrides.
Flash Point 91°C (195°F)
Quantity 100g
Melting Point 94°C to 98°C
Beilstein 2210199
Solubility Information Insoluble in water.
Formula Weight 156.12
Percent Purity 98%
Chemical Name or Material 4-Fluoro-3-nitroaniline

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 206-665-5

RTECSNumber : BY1700000

TSCA : Yes

Recommended Storage : Ambient temperatures