Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-3-nitrobenzotrifluoride, 96%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
Packaging:
Glass bottle
367-86-2
C7H3F4NO2
209.1
MFCD00007059
HLDFCCHSOZWKAA-UHFFFAOYSA-N
4-fluoro-3-nitrobenzotrifluoride,1-fluoro-2-nitro-4-trifluoromethyl benzene,benzene, 1-fluoro-2-nitro-4-trifluoromethyl,4-fluoro-3-nitrotrifluoromethylbenzene,3-nitro-4-fluorobenzotrifluoride,3-nitro-alpha,alpha,alpha,4-tetrafluorotoluene,2-fluoro-5-trifluoromethyl nitrobenzene,3-nitro-4-fluoro benzotrifluoride,1-fluoro-2-nitro-4-trifluoromethyl-benzene
67778
1-fluoro-2-nitro-4-(trifluoromethyl)benzene
C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])F
367-86-2
C7H3F4NO2
209.1
MFCD00007059
HLDFCCHSOZWKAA-UHFFFAOYSA-N
4-fluoro-3-nitrobenzotrifluoride,1-fluoro-2-nitro-4-trifluoromethyl benzene,benzene, 1-fluoro-2-nitro-4-trifluoromethyl,4-fluoro-3-nitrotrifluoromethylbenzene,3-nitro-4-fluorobenzotrifluoride,3-nitro-alpha,alpha,alpha,4-tetrafluorotoluene,2-fluoro-5-trifluoromethyl nitrobenzene,3-nitro-4-fluoro benzotrifluoride,1-fluoro-2-nitro-4-trifluoromethyl-benzene
67778
1-fluoro-2-nitro-4-(trifluoromethyl)benzene
C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])F
Boiling Point 92°C (15.0mmHg)
Flash Point 33°C
Density 1.4900g/mL
Refractive Index 1.4610 to 1.463
Specific Gravity 1.49
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5g
Color Yellow
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Linear Formula FC6H3(NO2)CF3
Beilstein 05, IV, 853
Formula Weight 209.1
Physical Form Liquid
Percent Purity 96%
Chemical Name or Material 4-Fluoro-3-nitrobenzotrifluoride
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 206-702-5