Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Fluoro-3-nitrotoluene, 98%

Click to view available options
Quantity:
25g
100g
446-11-7
C7H6FNO2
155.128
MFCD00007060
OORBDHOQLZRIQR-UHFFFAOYSA-N
4-fluoro-3-nitrotoluene,3-nitro-4-fluorotoluene,2-fluoro-5-methylnitrobenzene,4-methyl-2-nitro-1-fluorobenzene,1-fluoro-4-methyl-2-nitro-benzene,benzene, 1-fluoro-4-methyl-2-nitro,pubchem1597,acmc-1aqwi,4-fluro-3-nitrotoluene,4-fluoro-3-nitro toluene
136287
1-fluoro-4-methyl-2-nitrobenzene
CC1=CC(=C(C=C1)F)[N+](=O)[O-]
446-11-7
C7H6FNO2
155.128
MFCD00007060
OORBDHOQLZRIQR-UHFFFAOYSA-N
4-fluoro-3-nitrotoluene,3-nitro-4-fluorotoluene,2-fluoro-5-methylnitrobenzene,4-methyl-2-nitro-1-fluorobenzene,1-fluoro-4-methyl-2-nitro-benzene,benzene, 1-fluoro-4-methyl-2-nitro,pubchem1597,acmc-1aqwi,4-fluro-3-nitrotoluene,4-fluoro-3-nitro toluene
136287
1-fluoro-4-methyl-2-nitrobenzene
CC1=CC(=C(C=C1)F)[N+](=O)[O-]
Color Red to Brown
Boiling Point 241°C (decomposition)
Quantity 25g
UN Number UN2810
Beilstein 2253853
Formula Weight 155.13
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 4-Fluoro-3-nitrotoluene

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures