Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-3-(tetrazol-5-yl)phenylboronic acid, 95%, Acros Organics

Click to view available options
Quantity:
1g
5g
1009303-56-3
C7H6BFN4O2
207.96
FELFJHZMQYCKPT-UHFFFAOYSA-N
4-fluoro-3-tetrazol-5-yl phenylboronic acid,4-fluoro-3-1h-1,2,3,4-tetrazol-5-yl phenylboronic acid,4-fluoro-3-1h-tetrazol-5-yl phenyl boronic acid,4-fluoro-3-1h-tetrazol-5-yl phenylboronic acid
11287236
[4-fluoro-3-(2H-tetrazol-5-yl)phenyl]boronic acid
B(C1=CC(=C(C=C1)F)C2=NNN=N2)(O)O
1009303-56-3
C7H6BFN4O2
207.96
FELFJHZMQYCKPT-UHFFFAOYSA-N
4-fluoro-3-tetrazol-5-yl phenylboronic acid,4-fluoro-3-1h-1,2,3,4-tetrazol-5-yl phenylboronic acid,4-fluoro-3-1h-tetrazol-5-yl phenyl boronic acid,4-fluoro-3-1h-tetrazol-5-yl phenylboronic acid
11287236
[4-fluoro-3-(2H-tetrazol-5-yl)phenyl]boronic acid
B(C1=CC(=C(C=C1)F)C2=NNN=N2)(O)O
Quantity 5g
Assay Percent Range 94% min. (HPLC)
Formula Weight 207.96
Percent Purity 95%
Chemical Name or Material 4-Fluoro-3-(tetrazol-5-yl)phenylboronic acid

GHS Signal Word: Warning