Learn More
Alfa Aesar™ 4-Fluoro-3-(trifluoromethoxy)phenol, 97%
Manufacturer:  Alfa Aesar™ H26766MD
Quantity 250mg
 View more versions of this product
Catalog No. AAH26766MD
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
886501-26-4
196.1
4-Fluoro-3-(trifluoromethoxy)phenol
C7H4F4O2
MFCD06660257
Specifications
250mg
C7H4F4O2
MFCD06660257
97%
886501-26-4
196.1
4-Fluoro-3-(trifluoromethoxy)phenol
Safety and Handling
SDS & Safety Documents
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger