Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Fluoro-3-(trifluoromethyl)benzophenone, 97+%

Click to view available options
Quantity:
1g
5g
239087-04-8
C14H8F4O
268.211
MFCD00061258
BWBZTYREKMHBFH-UHFFFAOYSA-N
4-fluoro-3-trifluoromethyl benzophenone,4-fluoro-3-trifluoromethyl phenyl phenyl methanone,acmc-1cehg,4-fluoro-3-trifluoromethyl phenyl phenyl ketone,4-fluoro-3-trifluoromethyl phenyl-phenylmethanone,methanone, 4-fluoro-3-trifluoromethyl phenyl phenyl
2737556
[4-fluoro-3-(trifluoromethyl)phenyl]-phenylmethanone
C1=CC=C(C=C1)C(=O)C2=CC(=C(C=C2)F)C(F)(F)F
239087-04-8
C14H8F4O
268.211
MFCD00061258
BWBZTYREKMHBFH-UHFFFAOYSA-N
4-fluoro-3-trifluoromethyl benzophenone,4-fluoro-3-trifluoromethyl phenyl phenyl methanone,acmc-1cehg,4-fluoro-3-trifluoromethyl phenyl phenyl ketone,4-fluoro-3-trifluoromethyl phenyl-phenylmethanone,methanone, 4-fluoro-3-trifluoromethyl phenyl phenyl
2737556
[4-fluoro-3-(trifluoromethyl)phenyl]-phenylmethanone
C1=CC=C(C=C1)C(=O)C2=CC(=C(C=C2)F)C(F)(F)F
Quantity 5g
Melting Point 44°C to 47°C
Formula Weight 268.21
Percent Purity ≥97%
Chemical Name or Material 4-Fluoro-3-(trifluoromethyl)benzophenone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning