Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Fluoro-3-(trifluoromethyl)benzoyl chloride, 97%

Click to view available options
Quantity:
1g
5g
67515-56-4
C8H3ClF4O
226.555
MFCD00061159
BUDISZQHCHGLJW-UHFFFAOYSA-N
4-fluoro-3-trifluoromethyl benzoyl chloride,4-fluoro-3-trifluoromethyl benzoylchloride,4-fluoro-3-trifluoromethyl-benzoyl chloride,benzoyl chloride,4-fluoro-3-trifluoromethyl,benzoyl chloride, 4-fluoro-3-trifluoromethyl,alpha,alpha,alpha,4-tetrafluoro-m-toluoyl chloride,acmc-1b7wd,4-fluoro-5-trifluoromethylbenzoyl chloride,5-chlorocarbonyl-2-fluorobenzotrifluoride
144253
4-fluoro-3-(trifluoromethyl)benzoyl chloride
C1=CC(=C(C=C1C(=O)Cl)C(F)(F)F)F
67515-56-4
C8H3ClF4O
226.555
MFCD00061159
BUDISZQHCHGLJW-UHFFFAOYSA-N
4-fluoro-3-trifluoromethyl benzoyl chloride,4-fluoro-3-trifluoromethyl benzoylchloride,4-fluoro-3-trifluoromethyl-benzoyl chloride,benzoyl chloride,4-fluoro-3-trifluoromethyl,benzoyl chloride, 4-fluoro-3-trifluoromethyl,alpha,alpha,alpha,4-tetrafluoro-m-toluoyl chloride,acmc-1b7wd,4-fluoro-5-trifluoromethylbenzoyl chloride,5-chlorocarbonyl-2-fluorobenzotrifluoride
144253
4-fluoro-3-(trifluoromethyl)benzoyl chloride
C1=CC(=C(C=C1C(=O)Cl)C(F)(F)F)F
Sensitivity Moisture sensitive
Refractive Index 1.4685
Density 1.493
Quantity 1g
Flash Point 76°C (168°F)
Boiling Point 169°C to 171°C
UN Number UN3094
Beilstein 1968721
Formula Weight 226.56
Percent Purity 97%
Chemical Name or Material 4-Fluoro-3-(trifluoromethyl)benzoyl chloride
Show More Show Less

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures