Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-4′-(trans-4-propylcyclohexyl)biphenyl, TCI America™

Click to view available options
Quantity:
1g
5g
87260-24-0
C21H25F
296.429
MFCD11053438
DYBQDZYHNOGWIK-UHFFFAOYSA-N
4-fluoro-4'-trans-4-propylcyclohexyl-1,1'-biphenyl,trans-4-4-propylcyclohexyl-4'-fluorobiphenyl,1-fluoro-4-4-4-propylcyclohexyl phenyl benzene,4-fluoro-4'-1s,4r-4-propylcyclohexyl-1,1'-biphenyl,4-4-propylcyclohexyl-4'-fluorobiphenyl,4-fluoro-4'-trans-4-proptlcyclohexyl biphenyl,rans-4-4-propylcyclohexyl-4-fluorobiphenyl,trans-4-4-propylcyclohexyl-4-fluorobiphenyl,trans-4-fluoro-4'-4-n-propylcyclohexyl biphenyl,1,1'-biphenyl, 4-fluoro-4'-4-propylcyclohexyl
611152
1-fluoro-4-[4-(4-propylcyclohexyl)phenyl]benzene
CCCC1CCC(CC1)C2=CC=C(C=C2)C3=CC=C(C=C3)F
87260-24-0
C21H25F
296.429
MFCD11053438
DYBQDZYHNOGWIK-UHFFFAOYSA-N
4-fluoro-4'-trans-4-propylcyclohexyl-1,1'-biphenyl,trans-4-4-propylcyclohexyl-4'-fluorobiphenyl,1-fluoro-4-4-4-propylcyclohexyl phenyl benzene,4-fluoro-4'-1s,4r-4-propylcyclohexyl-1,1'-biphenyl,4-4-propylcyclohexyl-4'-fluorobiphenyl,4-fluoro-4'-trans-4-proptlcyclohexyl biphenyl,rans-4-4-propylcyclohexyl-4-fluorobiphenyl,trans-4-4-propylcyclohexyl-4-fluorobiphenyl,trans-4-fluoro-4'-4-n-propylcyclohexyl biphenyl,1,1'-biphenyl, 4-fluoro-4'-4-propylcyclohexyl
611152
1-fluoro-4-[4-(4-propylcyclohexyl)phenyl]benzene
CCCC1CCC(CC1)C2=CC=C(C=C2)C3=CC=C(C=C3)F
Color White
Melting Point 106°C
Quantity 5g
Formula Weight 296.43
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material 4-Fluoro-4′-(trans-4-propylcyclohexyl)biphenyl