Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Fluoro-beta-nitrostyrene, 98%

Click to view available options
Quantity:
5g
25g
100g
706-08-1
C8H6FNO2
167.139
MFCD00024824
VRFSQVFSQAYHRU-AATRIKPKSA-N
1-fluoro-4-2-nitrovinyl benzene,4-fluoro-beta-nitrostyrene,e-1-fluoro-4-2-nitrovinyl benzene,trans-p-fluoro-beta-nitrostyrene,p-fluoro-beta-nitrostyrene,styrene, p-fluoro-beta-nitro,1-4-fluorophenyl-2-nitroethylene,trans p-fluorophenyl-1 nitro-2 ethylene french,styrene, p-fluoro-beta-nitro-, e,1-fluoro-4-e-2-nitrovinyl benzene
5381680
1-fluoro-4-[(E)-2-nitroethenyl]benzene
C1=CC(=CC=C1C=C[N+](=O)[O-])F
706-08-1
C8H6FNO2
167.139
MFCD00024824
VRFSQVFSQAYHRU-AATRIKPKSA-N
1-fluoro-4-2-nitrovinyl benzene,4-fluoro-beta-nitrostyrene,e-1-fluoro-4-2-nitrovinyl benzene,trans-p-fluoro-beta-nitrostyrene,p-fluoro-beta-nitrostyrene,styrene, p-fluoro-beta-nitro,1-4-fluorophenyl-2-nitroethylene,trans p-fluorophenyl-1 nitro-2 ethylene french,styrene, p-fluoro-beta-nitro-, e,1-fluoro-4-e-2-nitrovinyl benzene
5381680
1-fluoro-4-[(E)-2-nitroethenyl]benzene
C1=CC(=CC=C1C=C[N+](=O)[O-])F
Applications
It is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store at 99-102°C. Incompatible with oxidizing agents.
Color Yellow
Quantity 25g
Melting Point 99°C to 102°C
UN Number UN2811
Beilstein 2208372
Formula Weight 167.14
Physical Form Crystalline Solid
Percent Purity 98%
Chemical Name or Material 4-Fluoro-beta-nitrostyrene

GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement
P280-P264-P305+P351+P338-P310-P301+P312-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : WL4800000

storageNote1 : 2° to 8°C