Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-beta-nitrostyrene, 98%, Thermo Scientific™

Catalog Number AAH6461306
Click to view available options
Quantity:
5 g
25 g
100 g
706-08-1
C8H6FNO2
167.14
MFCD00024824
VRFSQVFSQAYHRU-AATRIKPKSA-N
1-fluoro-4-2-nitrovinyl benzene, 4-fluoro-beta-nitrostyrene, e-1-fluoro-4-2-nitrovinyl benzene, trans-p-fluoro-beta-nitrostyrene, p-fluoro-beta-nitrostyrene, styrene, p-fluoro-beta-nitro, 1-4-fluorophenyl-2-nitroethylene, trans p-fluorophenyl-1 nitro-2 ethylene french, styrene, p-fluoro-beta-nitro-, e, 1-fluoro-4-e-2-nitrovinyl benzene
5381680
[O-][N+](=O)\C=C\C1=CC=C(F)C=C1
706-08-1
C8H6FNO2
167.14
MFCD00024824
VRFSQVFSQAYHRU-AATRIKPKSA-N
1-fluoro-4-2-nitrovinyl benzene, 4-fluoro-beta-nitrostyrene, e-1-fluoro-4-2-nitrovinyl benzene, trans-p-fluoro-beta-nitrostyrene, p-fluoro-beta-nitrostyrene, styrene, p-fluoro-beta-nitro, 1-4-fluorophenyl-2-nitroethylene, trans p-fluorophenyl-1 nitro-2 ethylene french, styrene, p-fluoro-beta-nitro-, e, 1-fluoro-4-e-2-nitrovinyl benzene
5381680
[O-][N+](=O)\C=C\C1=CC=C(F)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store at 99-102°C. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 99°C to 102°C
Color Yellow
Quantity 5 g
UN Number UN2811
Beilstein 2208372
Solubility Information Slightly soluble in water.
IUPAC Name 1-fluoro-4-[(1E)-2-nitroethenyl]benzene
Formula Weight 167.14
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material 4-Fluoro-beta-nitrostyrene
Show More Show Less
Hazard Category H302-H318
Hazard Statement GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 211-891-2
RTECSNumber WL4800000
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit