Alfa Aesar™ 4-Fluoro-N-methylaniline, 97%

Manufacturer: Alfa Aesar™ H5510506

Quantity 5g

 View more versions of this product

Catalog No. AAH5510506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 459-59-6
    Molecular Formula C7FH8N
    Formula Weight 125.15
    MDL Number MFCD00017942
    UN Number UN2810
    Chemical Name or Material 4-Fluoro-N-methylaniline

    Specifications

    Formula Weight 125.15
    Flash Point 76°C (169°F)
    Boiling Point 79°C (11mmHg)
    Quantity 5g
    Molecular Formula C7FH8N
    MDL Number MFCD00017942
    CAS 459-59-6
    UN Number UN2810
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H318-H302-H317-H335-H227-H315
    Causes serious eye damage.
    Harmful if swallowed.
    May cause an allergic skin reaction.
    May cause respiratory irritation.
    Combustible liquid.
    Causes skin irritation.
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
    Chemical Name or Material 4-Fluoro-N-methylaniline
    Assay 97%

    Description

    Applications
    4-Fluoro-N-methylaniline is used as a model compound to study the in vivo and in vitro biotransformation of secondary aromatic amines.

    Notes
    Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents , acids, acid chloride, acid anhydride and chloroformates.

    GHS H Statement
    H318-H302-H317-H335-H227-H315
    Causes serious eye damage.
    Harmful if swallowed.
    May cause an allergic skin reaction.
    May cause respiratory irritation.
    Combustible liquid.
    Causes skin irritation.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit