Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Fluoro-N-methylaniline, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
4-Fluoro-N-methylaniline is used as a model compound to study the biotransformation of secondary aromatic amines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Fluoro-N-methylaniline is used as a model compound to study the biotransformation of secondary aromatic amines.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents , acids, acid chloride, acid anhydride and chloroformates.
4-Fluoro-N-methylaniline is used as a model compound to study the biotransformation of secondary aromatic amines.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents , acids, acid chloride, acid anhydride and chloroformates.
Specifications
Density | 1.04 |
Boiling Point | 79°C (11 mmHg) |
Flash Point | 76°C (169°F) |
Refractive Index | 1.532 |
Quantity | 1 g |
UN Number | UN2810 |
Synonym | benzenamine, 4-fluoro-n-methyl, unii-88f75yis0f, n-methyl-4-fluoroaniline, acmc-1aem5, 4-fluoro-n-methyl aniline, 4-fluoro-n-methyl-aniline, 4-fluoro-n-methylbenzenamine, 4-fluorophenyl-methylamine, benzenamine,4-fluoro-n-methyl |
Formula Weight | 125.15 |
Percent Purity | 97% |
Chemical Name or Material | 4-Fluoro-N-methylaniline |
Safety and Handling
Hazard Category | H227-H302+H312+H332-H315-H319-H335 |
Hazard Statement | GHS H Statement H318-H302-H317-H335-H227-H315 Causes serious eye damage. Harmful if swallowed. May cause an allergic skin reaction. May cause respiratory irritation. Combustible liquid. Causes skin irritation. |
Precautionary Statement | P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S. |
EINECSNumber | 207-294-1 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction