Alfa Aesar™ 4-Fluoro-N-methylbenzylamine, 97%

Manufacturer: Alfa Aesar™ H54369-03

 View more versions of this product

Catalog No. AAH5436903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 405-66-3
    Molecular Formula C8H10FN
    Formula Weight 139.17g/mol
    MDL Number MFCD03839848
    Chemical Name or Material 4-Fluoro-N-methylbenzylamine

    Specifications

    Boiling Point 180° to 181°C
    Quantity 1g
    Flash Point 71°C (160°F)
    CAS 405-66-3
    Molecular Formula C8H10FN
    Formula Weight 139.17g/mol
    MDL Number MFCD03839848
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H318-H317-H335-H227-H315
    Causes serious eye damage.
    May cause an allergic skin reaction.
    May cause respiratory irritation.
    Combustible liquid.
    Causes skin irritation.
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 4-Fluoro-N-methylbenzylamine
    Assay 97%

    GHS H Statement
    H318-H317-H335-H227-H315
    Causes serious eye damage.
    May cause an allergic skin reaction.
    May cause respiratory irritation.
    Combustible liquid.
    Causes skin irritation.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit