Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-N-methylbenzylamine, 97%, Thermo Scientific Chemicals

Catalog Number AAH5436906
Click to view available options
Quantity:
1 g
5 g
405-66-3
C8H10FN
139.173
MFCD03839848
SZJIQLSCDIEJFC-UHFFFAOYSA-N
n-methyl-4-fluorobenzylamine, 4-fluoro-n-methylbenzylamine, 4-fluoro-benzyl-methyl-amine, 1-4-fluorophenyl-n-methylmethanamine, n-4-fluorobenzyl-n-methylamine, 4-fluoro-n-methyl benzylamine, 4-fluorophenyl methyl methyl amine, benzenemethanamine, 4-fluoro-n-methyl, 1-4-fluorophenyl-n-methyl-methanamine, acmc-1aek3
736860
1-(4-fluorophenyl)-N-methylmethanamine
CNCC1=CC=C(C=C1)F
405-66-3
C8H10FN
139.173
MFCD03839848
SZJIQLSCDIEJFC-UHFFFAOYSA-N
n-methyl-4-fluorobenzylamine, 4-fluoro-n-methylbenzylamine, 4-fluoro-benzyl-methyl-amine, 1-4-fluorophenyl-n-methylmethanamine, n-4-fluorobenzyl-n-methylamine, 4-fluoro-n-methyl benzylamine, 4-fluorophenyl methyl methyl amine, benzenemethanamine, 4-fluoro-n-methyl, 1-4-fluorophenyl-n-methyl-methanamine, acmc-1aek3
736860
1-(4-fluorophenyl)-N-methylmethanamine
CNCC1=CC=C(C=C1)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.056
Boiling Point 180°C to 181°C
Flash Point 71°C (160°F)
Refractive Index 1.499
Quantity 5 g
Sensitivity Air Sensitive
Formula Weight 139.17
Percent Purity 97%
Chemical Name or Material 4-Fluoro-N-methylbenzylamine
Hazard Category H227-H315-H318-H334-H335
Hazard Statement GHS H Statement
H318-H317-H335-H227-H315
Causes serious eye damage.
May cause an allergic skin reaction.
May cause respiratory irritation.
Combustible liquid.
Causes skin irritation.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P285-P302+P352-P304+P340-P305+P351+P338-P310-P332+P313-P342+P311-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit