Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Fluoro-N,N-dimethyl-2-nitroaniline, 97%

Click to view available options
Quantity:
250mg
1g
5g
69261-01-4
C8H9FN2O2
184.17
MFCD04116334
RGCUVPLVTHHXMT-UHFFFAOYSA-N
n,n-dimethyl 4-fluoro-2-nitroaniline,n,n-dimethyl-4-fluoro-2-nitroaniline,benzenamine,4-fluoro-n,n-dimethyl-2-nitro,pubchem3366,acmc-1b82t,4-fluoro-2-nitrophenyl dimethylamine,4-fluoranyl-n,n-dimethyl-2-nitro-aniline
16440018
4-fluoro-N,N-dimethyl-2-nitroaniline
CN(C)C1=C(C=C(C=C1)F)[N+](=O)[O-]
69261-01-4
C8H9FN2O2
184.17
MFCD04116334
RGCUVPLVTHHXMT-UHFFFAOYSA-N
n,n-dimethyl 4-fluoro-2-nitroaniline,n,n-dimethyl-4-fluoro-2-nitroaniline,benzenamine,4-fluoro-n,n-dimethyl-2-nitro,pubchem3366,acmc-1b82t,4-fluoro-2-nitrophenyl dimethylamine,4-fluoranyl-n,n-dimethyl-2-nitro-aniline
16440018
4-fluoro-N,N-dimethyl-2-nitroaniline
CN(C)C1=C(C=C(C=C1)F)[N+](=O)[O-]
Quantity 250mg
UN Number UN2811
Formula Weight 184.17
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 4-Fluoro-N,N-dimethyl-2-nitroaniline

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures