Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-o-xylene, 98%, Acros Organics

Click to view available options
Quantity:
25g
452-64-2
C8H9F
124.158
MFCD00039217
WYHBENDEZDFJNU-UHFFFAOYSA-N
4-fluoro-o-xylene,1,2-dimethyl-4-fluorobenzene,3,4-dimethylfluorobenzene,4-fluoro-1,2-xylene,1-fluoro-3,4-dimethylbenzene,benzene, 4-fluoro-1,2-dimethyl,o-xylene, 4-fluoro,pubchem14118,acmc-1afww,3,4-dimethyl fluorobenzene
253306
4-fluoro-1,2-dimethylbenzene
CC1=C(C=C(C=C1)F)C
452-64-2
C8H9F
124.158
MFCD00039217
WYHBENDEZDFJNU-UHFFFAOYSA-N
4-fluoro-o-xylene,1,2-dimethyl-4-fluorobenzene,3,4-dimethylfluorobenzene,4-fluoro-1,2-xylene,1-fluoro-3,4-dimethylbenzene,benzene, 4-fluoro-1,2-dimethyl,o-xylene, 4-fluoro,pubchem14118,acmc-1afww,3,4-dimethyl fluorobenzene
253306
4-fluoro-1,2-dimethylbenzene
CC1=C(C=C(C=C1)F)C
Color Colorless to Yellow
Refractive Index 1.4805 to 1.4825 (20°C, 589nm)
Quantity 25g
Infrared Spectrum Authentic
Density 1g/mL
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 124.16
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 4-Fluoro-o-xylene, 98%
Show More Show Less