Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Fluoroacetophenone, 99%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
500mL
Packaging:
Glass bottle
403-42-9
C8H7FO
138.14
MFCD00000354
ZDPAWHACYDRYIW-UHFFFAOYSA-N
4'-fluoroacetophenone,4-fluoroacetophenone,1-4-fluorophenyl ethanone,p-fluoroacetophenone,1-4-fluorophenyl ethan-1-one,ethanone, 1-4-fluorophenyl,acetophenone, 4'-fluoro,4 fluoroacetophenone,unii-tvq090602v,1-acetyl-4-fluorobenzene
9828
1-(4-fluorophenyl)ethanone
CC(=O)C1=CC=C(C=C1)F
403-42-9
C8H7FO
138.14
MFCD00000354
ZDPAWHACYDRYIW-UHFFFAOYSA-N
4'-fluoroacetophenone,4-fluoroacetophenone,1-4-fluorophenyl ethanone,p-fluoroacetophenone,1-4-fluorophenyl ethan-1-one,ethanone, 1-4-fluorophenyl,acetophenone, 4'-fluoro,4 fluoroacetophenone,unii-tvq090602v,1-acetyl-4-fluorobenzene
9828
1-(4-fluorophenyl)ethanone
CC(=O)C1=CC=C(C=C1)F
Melting Point 4°C
Boiling Point 196°C
Flash Point 71°C
Specific Gravity 1.13
Refractive Index 1.5090 to 1.512
Color Colorless to Yellow
Infrared Spectrum Authentic
Quantity 25mL
Density 1.1300g/mL
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula FC6H4COCH3
Formula Weight 138.14
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material 4'-Fluoroacetophenone, 99%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 206-960-9

TSCA : TSCA