Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluorobenzamidoxime, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
Packaging:
Glass bottle
69113-32-2
C7H7FN2O
154.15
MFCD00216571
OSUPWUQRPLIJKX-UHFFFAOYSA-N
4-fluoro-n-hydroxybenzimidamide,4-fluoro-n'-hydroxybenzimidamide,z-4-fluoro-n'-hydroxybenzenecarboximidamide,4-fluorophenyl hydroxyimino methylamine,benzenecarboximidamide,4-fluoro-n-hydroxy,p-fluorobenzamidoxime,#,zyb
5960802
4-fluoro-N'-hydroxybenzenecarboximidamide
C1=CC(=CC=C1C(=NO)N)F
69113-32-2
C7H7FN2O
154.15
MFCD00216571
OSUPWUQRPLIJKX-UHFFFAOYSA-N
4-fluoro-n-hydroxybenzimidamide,4-fluoro-n'-hydroxybenzimidamide,z-4-fluoro-n'-hydroxybenzenecarboximidamide,4-fluorophenyl hydroxyimino methylamine,benzenecarboximidamide,4-fluoro-n-hydroxy,p-fluorobenzamidoxime,#,zyb
5960802
4-fluoro-N'-hydroxybenzenecarboximidamide
C1=CC(=CC=C1C(=NO)N)F
Melting Point 95.2°C to 96.1°C
Color White
Quantity 10g
Packaging Glass bottle
Infrared Spectrum Authentic
Elemental Analysis 53.18 to 55.91% C, 17.72 to 18.63% N
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 154.15
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 4-Fluorobenzamidoxime
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning