Alfa Aesar™ 4-Fluorobenzenesulfonamide, 98+%

Manufacturer: Alfa Aesar™ H5583406

Quantity 5g

 View more versions of this product

Catalog No. AAH5583406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 402-46-0
    Molecular Formula C6H6FNO2S
    Formula Weight 175.18
    MDL Number MFCD00025384
    UN Number UN2811
    Chemical Name or Material 4-Fluorobenzenesulfonamide

    Specifications

    Formula Weight 175.18
    Molecular Formula C6H6FNO2S
    MDL Number MFCD00025384
    UN Number UN2811
    Health Hazard 1 Warning
    RTECS Number DB2630000
    Melting Point 124° to 127°C
    Quantity 5g
    CAS 402-46-0
    Health Hazard 2 GHS H Statement
    H302-H312-H332-H315-H319-H335
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Chemical Name or Material 4-Fluorobenzenesulfonamide
    Assay ≥98%

    Description

    Applications
    4-Fluorobenzenesulfonamide, is used to produce bis-(4-fluoro-benzenesulfonyl)-amine with 4-fluoro-benzenesulfonyl chloride at temperature of 55 - 60. This reaction will need reagent OH-.

    Solubility
    It is soluble in water.

    Notes
    Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

    GHS H Statement
    H302-H312-H332-H315-H319-H335
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit