Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluorobenzenesulfonyl chloride, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
349-88-2
C6H4ClFO2S
194.61
MFCD00007438
BFXHJFKKRGVUMU-UHFFFAOYSA-N
4-fluorobenzene-1-sulfonyl chloride,p-fluorobenzenesulfonyl chloride,4-fluorobenzenesulphonyl chloride,benzenesulfonyl chloride, 4-fluoro,4-fluorophenylsulfonyl chloride,4-fluorobenzenesulfonic acid chloride,4-fluorobenzenesulfonylchloride,benzenesulfonyl chloride, p-fluoro,4-fluorobenzensulfonyl chloride,4-fluoro-benzenesulfonyl chloride
9588
4-fluorobenzenesulfonyl chloride
C1=CC(=CC=C1F)S(=O)(=O)Cl
349-88-2
C6H4ClFO2S
194.61
MFCD00007438
BFXHJFKKRGVUMU-UHFFFAOYSA-N
4-fluorobenzene-1-sulfonyl chloride,p-fluorobenzenesulfonyl chloride,4-fluorobenzenesulphonyl chloride,benzenesulfonyl chloride, 4-fluoro,4-fluorophenylsulfonyl chloride,4-fluorobenzenesulfonic acid chloride,4-fluorobenzenesulfonylchloride,benzenesulfonyl chloride, p-fluoro,4-fluorobenzensulfonyl chloride,4-fluoro-benzenesulfonyl chloride
9588
4-fluorobenzenesulfonyl chloride
C1=CC(=CC=C1F)S(=O)(=O)Cl
Melting Point 29°C to 34°C
Flash Point >110°C
Boiling Point 95°C to 96°C (2.0mmHg)
Packaging Glass bottle
Color Brown to White
Infrared Spectrum Authentic
Quantity 25g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula FC6H4SO2Cl
Beilstein 11, 53
Formula Weight 194.61
Physical Form Crystalline Low Melting Mass
Percent Purity ≥97.5% (ex Cl)
Chemical Name or Material 4-Fluorobenzenesulfonyl chloride
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger