Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Fluorobenzyl alcohol, 97%

Manufacturer:  Alfa Aesar™ A1233606

Quantity 5g

 View more versions of this product

Catalog No. AAA1233606


  Check Availability
Add to cart

Specifications

Description

Applications
4-Fluorobenzyl alcohol is used in the preparation of many compounds including amides. It is also used as an pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and acid anhydrides.
Description & Specifications

Chemical Identifiers

459-56-3
126.13
1446930
C7H7FO
MFCD00004651
4-Fluorobenzyl alcohol

Specifications

90°C (194°F)
206°C
5g
C7H7FO
MFCD00004651
4-Fluorobenzyl alcohol
Characteristic
22° to 24°C
459-56-3
126.13
1446930
97%

Safety and Handling

SDS & Safety Documents

Warning

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit