Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Fluorobenzyl mercaptan, 96%

Manufacturer:  Alfa Aesar™ A1012603

Quantity 1g

 View more versions of this product

Catalog No. AAA1012603


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

15894-04-9
142.2
UN2810
4-Fluoro-alpha-toluenethiol
C7H7FS
MFCD00040724
2242520
4-Fluorobenzyl mercaptan

Specifications

76°C (168°F)
1g
C7H7FS
MFCD00040724
2242520
4-Fluorobenzyl mercaptan
72° to 74°C (15mmHg)
15894-04-9
142.2
UN2810
4-Fluoro-alpha-toluenethiol
96%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit